Fisherbrand™ Traceable™ Handheld Digital Barometer
Keep track of your environment and your time with this multi-functional Traceable™ Handheld Digital Barometer.
 View more versions of this product
Catalog No. 02-406
Explore available promotions
Description
Description
Keep track of your environment and your time with this multi-functional Traceable™ Handheld Digital Barometer.
  • Stopwatch/time of day clock
  • Barometric pressure trend displays current reading and previous 2, 4, 6, 8, 10 and 12 hours in a bar chart format
  • Minimum/maximum memory for altitudes reached during any time period
  • Barometer Accuracy: ±8 mbar (0.2363 in. Hg)
Certifications
Traceable to standards provided by NIST™. Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA.
Specifications
Specifications
-10°C to +60°C
0.1°C
Barometer, Lanyard, Two Button-cell Batteries, Shock-resistant Case, Battery
±8 mbar
14°F to 140°F
Handheld Digital Barometer