Fisherbrand™ Traceable™ Hi-Accuracy Thermometer
Traceable™ Hi-Accuracy Thermometer provides the exact readings required for critical requirements, quality control checks, and routine measurements.
 View more versions of this product
Catalog No. 15-078-250
Explore available promotions
Description
Description
Traceable™ Hi-Accuracy Thermometer provides the exact readings required for critical requirements, quality control checks, and routine measurements.
  • Available with one probe or two probes
  • Monitor inlet and outlet simultaneously
  • Visual and audible, high and low settable alarms
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
Specifications
ISO 17025, A2LA, NIST
3.05 mm
Measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater
10.79 cm
60 in.
226 mm
15.24 cm
5 oz.
2.75 in.
9 in.
70°C
-50°C
-50°C to +70°C
±0.3°C
6.98 cm
0.12 in.
9 x 2.75 x 4.25 in. (22.86 x 6.98 x 10.79 cm)
4.25 in.
Stainless Steel Probe, Traceable Certificate and Two AA Batteries
6.25 in.
152.4 cm
0.01°C
141.7 g
Traceable Thermometer
158°F
-58°F
-58°F to +158°F
AA
LED