missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Traceable™ Hi-Accuracy Thermometer
Traceable™ Hi-Accuracy Thermometer provides the exact readings required for critical requirements, quality control checks, and routine measurements.
$326.59 - $400.33
Specifications
| Certifications/Compliance | ISO 17025, A2LA, NIST |
|---|---|
| Diameter (English) Probe | 0.12 in. |
| Diameter (Metric) Probe | 3.05 mm |
| Dimensions (L x W x H) | 9 x 2.75 x 4.25 in. (22.86 x 6.98 x 10.79 cm) |
| For Use With (Application) | Measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater |
Description
Traceable™ Hi-Accuracy Thermometer provides the exact readings required for critical requirements, quality control checks, and routine measurements.
- Available with one probe or two probes
- Monitor inlet and outlet simultaneously
- Visual and audible, high and low settable alarms
- Includes Traceable Certificate and 2 AA Batteries
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
| ISO 17025, A2LA, NIST | |
| 3.05 mm | |
| Measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater | |
| 10.79 cm | |
| 6.25 in. | |
| 152.4 cm | |
| 0.01°C | |
| 141.7 g | |
| Traceable Thermometer | |
| 158°F | |
| -58°F | |
| -58°F to +158°F | |
| Yes | |
| ±0.3°C | |
| 6.98 cm |
| 0.12 in. | |
| 9 x 2.75 x 4.25 in. (22.86 x 6.98 x 10.79 cm) | |
| 4.25 in. | |
| 60 in. | |
| 226 mm | |
| 15.24 cm | |
| 5 oz. | |
| 2.75 in. | |
| 9 in. | |
| 70°C | |
| -50°C | |
| -50°C to +70°C | |
| AA | |
| LED |