Fisherbrand™ Traceable™ Hi-Accuracy Thermometer
Traceable™ Hi-Accuracy Thermometer provides the exact readings required for critical requirements, quality control checks, and routine measurements.
 View more versions of this product
Catalog No. 15-078-251
Explore available promotions
Description
Description
Traceable™ Hi-Accuracy Thermometer provides the exact readings required for critical requirements, quality control checks, and routine measurements.
  • Available with one probe or two probes
  • Monitor inlet and outlet simultaneously
  • Visual and audible, high and low settable alarms
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
Specifications
0.12 in.
9 x 2.75 x 4.25 in. (22.86 x 6.98 x 10.79 cm)
4.25 in.
Two Stainless Steel Probes, Traceable Certificate, and Two AA Batteries
6.25 in.
152.4 cm
Stainless Steel
5 oz.
2.75 in.
9 in.
70°C
-50°C
-50°C to +70°C
±0.3°C
6.98 cm
3.05 mm
Measuring temperatures in water baths, cuvets, gas systems, reagents, incubators, pharmaceuticals, petroleum products, foods, wastewater
10.79 cm
60 in.
226 mm
15.24 cm
0.01°C
141.7 g
Traceable Thermometer
158°F
-58°F
-58°F to +158°F
AA
LED