missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™
IUPAC formulation
Description
IUPAC formulation
- Compatible with all makes and instruments and electrodes
- Accuracy at 25°C is ±0.010 pH
Specifications
pH Buffer Standards | |
Instruments and Electrodes | |
Liquid | |
Buffer | |
Bottle | |
Certified |
Calibration | |
4.005 | |
16 oz. | |
Accuracy at 25°C is ±0010pH | |
Traceable to NIST |