missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™
IUPAC formulation
$53.65 - $56.26
Specifications
Chemical Name or Material | pH Buffer Standards |
---|---|
For Use With (Application) | Calibration |
For Use With (Equipment) | Instruments and Electrodes |
Physical Form | Liquid |
Solution Type | Buffer |
Catalog Number | Mfr. No. | Quantity | Packaging | pH | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | pH | Price | Quantity | ||||
06-664-262
|
N/A | 6 x 100 mL | 6 shots of 100mL | 4.005 |
Pack of 6 Each for $56.26
|
|
||||
06-664-259
|
N/A | 16 oz. | Bottle | 4.005 |
Each for $53.65
|
|
||||
06-664-263
|
N/A | 6 x 100 mL | 6 shots of 100mL | 7.00 |
Pack of 6 Each for $56.26
|
|
||||
06-664-260
|
N/A | 16 oz. | Bottle | 7.0 |
Each for $53.65
|
|
||||
06-664-264
|
N/A | 6 x 100 mL | 6 shots of 100mL | 10.012 |
Pack of 6 Each for $56.26
|
|
||||
06-664-261
|
N/A | 16 oz. | Bottle | 10.012 |
Each for $53.65
|
|
||||
Description
IUPAC formulation
- Compatible with all makes and instruments and electrodes
- Accuracy at 25°C is ±0.010 pH
Specifications
pH Buffer Standards | |
Instruments and Electrodes | |
Buffer | |
Traceable to NIST |
Calibration | |
Liquid | |
Accuracy at 25°C is ±0010pH | |
Certified |