missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™

IUPAC formulation

 View more versions of this product

Catalog No. 06-664-261


Explore available promotions

Description

Description

IUPAC formulation

  • Compatible with all makes and instruments and electrodes
  • Accuracy at 25°C is ±0.010 pH
Specifications

Specifications

pH Buffer Standards
10.012
Buffer
Bottle
Certified
Instruments and Electrodes
16 oz.
Accuracy at 25°C is ±0010pH
Traceable to NIST
Videos
SDS
Documents

Documents

Promotions

Promotions