missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Tubing Connectors and Adapters

Work in any tubing or vacuum system.

$42.62 - $64.03

Specifications

Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Used for any tubing or vacuum system
Color White
Maximum Pressure 20 psi
View More Specs

Products 3
Catalog Number Mfr. No. For Use With Overall Length (Metric) Shape Price Quantity  
Catalog Number Mfr. No. For Use With Overall Length (Metric) Shape Price Quantity  
S50708
View Documents
Fisherbrand™
227546EMD
3 to 7.8mm 85 mm Straight-stepped
Pack of 10 Each for $51.33
Case of 10 Pack for $513.30
Only null left
 
S50709
View Documents
Fisherbrand™
227555
3 to 7.8mm 83.82 mm Straight-stepped
Pack of 10 Each for $64.03
Case of 10 Pack for $640.30
Only null left
 
S50700B
View Documents
Fisherbrand™
EPD227245EMD
3 to 5.3mm 36.8 mm Straight
Pack of 20 Each for $42.62
Only null left
 
Description

Description

  • Excellent chemical and temperature resistance
  • Serrated connecting surfaces to prevent tubing slippage
  • Autoclavable Polypropylene
  • Adapters allow for connecting different diameter tubing
Specifications

Specifications

Autoclavable
Used for any tubing or vacuum system
20 psi
Polypropylene
White
Tubing Connector
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'