missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs.
Easy to fill and ready to use
Supplier: Fisherbrand™ 3024020504
Includes: Come with colored polypropylene closures with PTFE membrane (except 02-897-2, which has an HDPE closure).
Description
Easy to fill and ready to use
- Wide mouth
- Translucent and sputter-free
- With easy-to-read color-coded silk-screened labels
- Tips can be cut back to increase flow
- When not in use, bottles relieve internal pressure without dripping
Label information includes:
- Chemical name
- National Fire Protection Association (NFPA) codes and symbols
- Chemical Abstracts Service (CAS) number
- Hazard codes
- Laboratory protective equipment information
- Target organ information
- Recommended fill level
- LDPE recycling code
- Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement
Not recommended for use above 60°C (140°F).
Order Info
Available in cases of six, all with the same label; in a six-bottle Solvent Pack (one each labeled acetone, deionized water, ethanol, isopropanol, methanol and universal); or six-bottle Cleaning Pack (one each labeled acetone, bleach, deionized water, saline, soap and universal bottles).
Specifications
17 oz., 500mL | |
17 oz. | |
2.99 in. | |
LDPE | |
ISO 13485:2003 | |
Not Autoclavable | |
Wide | |
173 mm | |
6.81 in. | |
Polypropylene | |
76 mm | |
No |
500 mL | |
Blue | |
Color-coded Silk screened labels, PTFE membrane | |
Blue | |
Yes | |
No | |
Cylindrical | |
Wash Bottle | |
Distilled Water | |
38-415 mm | |
Polypropylene Closure |