missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs.
Easy to fill and ready to use
$110.88 - $180.29
Specifications
Capacity | 17 oz., 500mL |
---|---|
Capacity (Metric) | 500 mL |
Capacity (English) | 17 oz. |
Diameter (English) | 2.99 in. |
Material | LDPE |
Catalog Number | Mfr. No. | Color | For Use With (Application) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | For Use With (Application) | Price | Quantity | ||||
02-897-12
|
Fisherbrand™
3024020515 |
Assorted | Solvent assortment pack |
Case of 6 Each for $178.88
|
|
||||
02-897-13
|
Fisherbrand™
3024020520 |
Assorted | Cleaning assortment pack |
Case of 6 Each for $180.29
|
|
||||
02-897-3
|
Fisherbrand™
3024020503 |
Blue | Deionized Water |
Case of 6 Each for $118.12
|
|
||||
02-897-9
|
Fisherbrand™
3024020509 |
Blue | Soap |
Case of 6 Each for $120.97
|
|
||||
02-897-11
|
Fisherbrand™
3024020511 |
Blue | Water |
Case of 6 Each for $113.59
|
|
||||
02-897-4
|
Fisherbrand™
3024020504 |
Blue | Distilled Water |
Case of 6 Each for $113.59
|
|
||||
02-897-7
|
Fisherbrand™
3024020507 |
Green | Methanol |
Case of 6 Each for $113.59
|
|
||||
02-897-8
|
Fisherbrand™
3024020508 |
Natural | Saline |
Case of 6 Each for $120.97
|
|
||||
02-897-10
|
Fisherbrand™
3024020510 |
Natural | Universal |
Case of 6 Each for $110.88
|
|
||||
02-897-2
|
Fisherbrand™
3024020502 |
Natural | Bleach |
Case of 6 Each for $120.97
|
|
||||
02-897-5
|
Fisherbrand™
3024020505 |
Natural | Ethanol |
Case of 6 Each for $113.59
|
|
||||
02-897-1
|
Fisherbrand™
3024020501 |
Red | Acetone |
Case of 6 Each for $113.59
|
|
||||
02-897-6
|
Fisherbrand™
3024020506 |
Yellow | Isopropanol |
Case of 6 Each for $122.18
|
|
||||
Description
Easy to fill and ready to use
- Wide mouth
- Translucent and sputter-free
- With easy-to-read color-coded silk-screened labels
- Tips can be cut back to increase flow
- When not in use, bottles relieve internal pressure without dripping
Label information includes:
- Chemical name
- National Fire Protection Association (NFPA) codes and symbols
- Chemical Abstracts Service (CAS) number
- Hazard codes
- Laboratory protective equipment information
- Target organ information
- Recommended fill level
- LDPE recycling code
- Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement
Not recommended for use above 60°C (140°F).
Order Info
Available in cases of six, all with the same label; in a six-bottle Solvent Pack (one each labeled acetone, deionized water, ethanol, isopropanol, methanol and universal); or six-bottle Cleaning Pack (one each labeled acetone, bleach, deionized water, saline, soap and universal bottles).
Specifications
17 oz., 500mL | |
17 oz. | |
LDPE | |
Not Autoclavable | |
Wide | |
173 mm | |
6.81 in. | |
76 mm |
500 mL | |
2.99 in. | |
Yes | |
No | |
Cylindrical | |
Wash Bottle | |
38-415 mm | |
No |