missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs.

Easy to fill and ready to use

Supplier:  Fisherbrand™ 3024020505

 View more versions of this product

Catalog No. 02-897-5


Explore available promotions

Includes: Come with colored polypropylene closures with PTFE membrane (except 02-897-2, which has an HDPE closure).

Description

Description

Easy to fill and ready to use

  • Wide mouth
  • Translucent and sputter-free
  • With easy-to-read color-coded silk-screened labels
  • Tips can be cut back to increase flow
  • When not in use, bottles relieve internal pressure without dripping

Label information includes:

  • Chemical name
  • National Fire Protection Association (NFPA) codes and symbols
  • Chemical Abstracts Service (CAS) number
  • Hazard codes
  • Laboratory protective equipment information
  • Target organ information
  • Recommended fill level
  • LDPE recycling code
  • Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement

Not recommended for use above 60°C (140°F).

Order Info

Available in cases of six, all with the same label; in a six-bottle Solvent Pack (one each labeled acetone, deionized water, ethanol, isopropanol, methanol and universal); or six-bottle Cleaning Pack (one each labeled acetone, bleach, deionized water, saline, soap and universal bottles).

Specifications

Specifications

17 oz., 500mL
17 oz.
2.99 in.
LDPE
ISO 13485:2003
Not Autoclavable
Wide
173 mm
6.81 in.
Polypropylene
76 mm
No
500 mL
Natural
Color-coded Silk screened labels, PTFE membrane
Natural
Yes
No
Cylindrical
Wash Bottle
Ethanol
38-415 mm
Polypropylene Closure
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions