Fisherbrand™ 96-Well Polystyrene Plates
Ideal for antibiotic screens, cell-based assays and screening compounds.
 View more versions of this product
Catalog No. 12-565-501
Explore available promotions
Description
Description
Ideal for antibiotic screens, cell-based assays and screening compounds
Designed for compatibility with automated equipment.
  • Polystyrene
  • Plates stack easily without lids
  • Available in flat or round well shapes
  • Nonsterile
  • Untreated, provides uniform surface properties
  • Lid is sold separately (12-565-511)
  • Cat No 12-565-501 is Clear in color
Specifications
Specifications
Standard
8 x 12
Clear
Polystyrene
96
Non-sterile
Flat Bottom (F-well)
80/Cs.
Without Lid
80 Cases
Microplate
400 μL
 50-250 μL/well
Non-treated
Ideal for antibiotic screens, cell based assays and screening compounds
Product Suggestions
Product Suggestions