missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ 96-Well Polystyrene Plates
Ideal for antibiotic screens, cell-based assays and screening compounds.
$566.66 - $2217.32
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Material | Polystyrene |
No. of Wells | 96 |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Color | Well Shape | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Well Shape | Price | Quantity | |||||
12-565-501
|
N/A | Clear | Flat Bottom (F-well) |
Case of 80 Each for $566.66
|
|
|||||
12-565-500
|
N/A | Clear | Round Bottom (U-well) |
Case of 80 Each for $579.62
|
|
|||||
12-566-619
|
Fisherbrand™
236113 |
White | Flat Bottom (F-well) |
Case of 180 Each for $2,217.32
|
|
|||||
12-566-620
|
Fisherbrand™
237115 |
Black | Flat Bottom (F-well) |
Case of 180 Each for $2,128.68
|
|
|||||
Description
Ideal for antibiotic screens, cell-based assays and screening compounds
Designed for compatibility with automated equipment
- Polystyrene
- Plates stack easily without lids
- Available in flat or round well shapes
- Nonsterile
- Untreated, provides uniform surface properties
- Use 12-565-511 lid (sold separately)
Specifications
Standard | |
Polystyrene | |
Non-sterile | |
Ideal for antibiotic screens, cell based assays and screening compounds |
8 x 12 | |
96 | |
Non-treated |