Fisherbrand™ Absorbent Underpads
Quickly absorb liquids and keep spills from spreading
 View more versions of this product
Catalog No. 14-206-62
Explore available promotions
Description
Description
Absorb liquids quickly and contain spills
Cushioned surface minimizes breakage of delicate labware
  • Made with high-absorbency fibers
  • Polyethylene backing is chemical resistant, and the spunbond top layer provides durability even when wet
  • Lie flat, hugging the benchtop to prevent sliding
  • Absorb up to 750mL/m2
Specifications
Specifications
Absorbent Underpad
750 mL/m2
610 mm
510 mm
Quickly absorb liquids and keep spills from spreading
Polyethylene
24 in.
20 in.
24 x 20 in. (610 x 510mm)