Fisherbrand™ Acid/Alkali Test Papers
Foolproof pH test to determine if a solution is acidic or alkaline.
Description
- A foolproof pH test to determine if a solution is acidic
- The Blue Litmus test paper does not come with a color chart as it cannot provide you with a defined pH value
- The test strip will turn red if dipped into an acidic solution (pH <7)
- It will remain unchanged in an alkaline solution
- We have tested the blue litmus with multiple pH buffers
- The blue paper is blue at pH 8
- At pH 7, the sample is maybe just a little red
- The real change to red seems to start with pH 6
- 5 and lower
- Neutral Litmus test paper tests for Neutrality of a solution, or its acidity or alkalinity
- The test strip will turn red if dipped into an acidic solution (pH <7) or blue if dipped into an alkaline solution (pH >7)
- Red Litmus test paper is a foolproof pH test to determine if a solution is alkaline
- The test strip will turn blue if dipped into an alkaline solution (pH >7)
- Color chart not included
Order Info
100 Strips per vial
Specifications
OSHA, 29 CFR 1910.133, EN166 | |
0 to 14 | |
24 x 100 Strips | |
Litmus Neutral Test Strip |