Fisherbrand™ Acid/Alkali Test Papers
Foolproof pH test to determine if a solution is acidic or alkaline.
$86.00 - $149.62
Specifications
Certifications/Compliance | OSHA, 29 CFR 1910.133, EN166 |
---|---|
Quantity | 24 x 100 Strips |
Catalog Number | Mfr. No. | Type | Quantity | pH Range | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | pH Range | Price | Quantity | |||
14-845
|
N/A | Litmus Red Test Strip | 24 x 100 Strips | 7 to 14 |
Pack of 24 Each for $103.86
|
|
|||
14-861
|
N/A | Congo Red Test Strip | 24 x 100 Strips | Less than 3, blue; Greater than 5.2, red |
Pack of 24 Each for $149.62
|
|
|||
14-850
|
N/A | Litmus Neutral Test Strip | 24 x 100 Strips | 0 to 14 |
Pack of 24 Each for $104.38
|
|
|||
14-844
|
N/A | Blue Litmus Test Strip | 24 x 100 Strips | 0 to 7 |
Pack of 24 Each for $86.00
|
|
|||
Description
- A foolproof pH test to determine if a solution is acidic
- The Blue Litmus test paper does not come with a color chart as it cannot provide you with a defined pH value
- The test strip will turn red if dipped into an acidic solution (pH <7)
- It will remain unchanged in an alkaline solution
- We have tested the blue litmus with multiple pH buffers
- The blue paper is blue at pH 8
- At pH 7, the sample is maybe just a little red
- The real change to red seems to start with pH 6
- 5 and lower
- Neutral Litmus test paper tests for Neutrality of a solution, or its acidity or alkalinity
- The test strip will turn red if dipped into an acidic solution (pH <7) or blue if dipped into an alkaline solution (pH >7)
- Red Litmus test paper is a foolproof pH test to determine if a solution is alkaline
- The test strip will turn blue if dipped into an alkaline solution (pH >7)
- Color chart not included
Order Info
100 Strips per vial
Specifications
OSHA, 29 CFR 1910.133, EN166 | |
24 x 100 Strips |