missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Micro Stir Bars

Stirring small volumes in vessels such as vials, tubes and grandient makers.

Supplier:  Fisherbrand™ 371215011

 View more versions of this product

Catalog No. 16800522



Explore available promotions

Description

Description

  • Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
  • Available in a variety of colors and sizes
  • Alnico magnet encapsulated in USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Specifications

Specifications

2 mm
PTFE
Environmental testing and life science applications
Yellow
Autoclavable
Cylinder
0.197 in.
0.079 in.
Stir Bar
Alnico Magnet
5 mm
Videos
SDS
Documents

Documents

Promotions

Promotions