Fisherbrand™ Micro Stir Bars
Stirring small volumes in vessels such as vials, tubes and grandient makers.
$5.23 - $11.39
Catalog Number | Mfr. No. | Length (Metric) | Color | Diameter (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Color | Diameter (Metric) | Price | Quantity | |||
14-512-150
|
N/A | 8 mm | Blue | 1.5 mm |
Each for $6.59
Case of 50 Each for $329.50
|
|
|||
14-512-151
|
N/A | 15 mm | Blue | 1.5 mm |
Each for $6.76
Case of 50 Each for $338.00
|
|
|||
14-512-152
|
N/A | 2 mm | Blue | 2 mm |
Each for $9.00
Case of 50 Each for $450.00
|
|
|||
14-512-153
|
N/A | 5 mm | Blue | 2 mm |
Each for $11.39
Case of 50 Each for $569.50
|
|
|||
14-512-154
|
N/A | 7 mm | Blue | 2 mm |
Each for $9.14
Case of 50 Each for $457.00
|
|
|||
14-512-155
|
N/A | 3 mm | Blue | 3 mm |
Each for $8.03
Case of 50 Each for $401.50
|
|
|||
14-512-156
|
N/A | 10 mm | Blue | 3 mm |
Each for $10.46
Case of 50 Each for $523.00
|
|
|||
16800521
|
Fisherbrand™
371215010 |
5 mm | Red | 2 mm |
Each for $5.23
Case of 50 Each for $261.50
|
|
|||
16800522
|
Fisherbrand™
371215011 |
5 mm | Yellow | 2 mm |
Each for $5.23
Case of 50 Each for $261.50
|
|
|||
16800523
|
Fisherbrand™
371215013 |
7 mm | Red | 2 mm |
Each for $5.23
Case of 50 Each for $261.50
|
|
|||
16800524
|
Fisherbrand™
371215014 |
7 mm | Yellow | 2 mm |
Each for $5.23
Case of 50 Each for $261.50
|
|
|||
16800525
|
Fisherbrand™
371215001 |
8 mm | Red | 1.5 mm |
Each for $5.23
Case of 50 Each for $261.50
|
|
|||
16800526
|
Fisherbrand™
371215002 |
8 mm | Yellow | 1.5 mm |
Each for $5.23
Case of 50 Each for $261.50
|
|
|||
16800527
|
Fisherbrand™
371215022 |
10 mm | Red | 3 mm |
Each for $5.83
Case of 50 Each for $291.50
|
|
|||
16800528
|
Fisherbrand™
371215023 |
10 mm | Yellow | 3 mm |
Each for $5.83
Case of 50 Each for $291.50
|
|
|||
Description
- Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
- Available in a variety of sizes
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclave safe
Specifications
Autoclavable |