Fisherbrand™ Cell Scrapers
Easily harvest cells from tissue culture flasks
 View more versions of this product
Catalog No. 08-100-242
Explore available promotions
Description
Description
Easily harvest cells from tissue culture flasks
Fisherbrand Cell Scrapers are appropriate for a variety of cell culture vessels. The uniquely designed handle and blade reduce cell damage during the harvesting process.
  • Presterilized and individually wrapped in peel-open packaging
  • Nonpyrogenic
  • Polyethylene copolymer blade pivots to permit even contact with growth surface
  • Ribbed polystyrene handle provides secure grip and added control
  • Flexible joint between blade and handle eases entry into vessel
  • Pivoting blade improves position control to adjust for difficult angles during cell harvesting
Specifications
Specifications
1.18 in.
For a variety of cell culture vessels
39 cm
Cell Scraper
Polystyrene (Handle)
3 cm
15 in.
Polyethylene Copolymer
Large