Fisherbrand™ Clear Plastic Wrap
Designed to maintain sample purity.
 View more versions of this product
Catalog No. 22-305654
Description
Description
Designed specifically for the laboratory
  • Made of 100% ultraclean polyethylene—ensures sample purity and eliminates contamination
  • High-cling properties make it ideal for wrapping of beakers, flasks, or Petri dishes with an extraordinarily tight seal that stays in place
  • Low-tangle properties ensure wrap won't ball up when being handled
  • Carton has convenient metal tear-off bar
Specifications
Specifications
Sealing, Weighing, Transferring Samples
Clear
12 in.
30.4 m
100 ft.
Single Roll of Wrap
30cm
Polyethylene