Fisherbrand™ Compartmentalized Petri Dishes
Select from 2, 3, or 4-section plates with numbered sections
 View more versions of this product
Catalog No. FB08757150
Explore available promotions
Description
Description
Select from 2, 3, or 4-section plates with numbered sections
  • All dishes manufactured from virgin medical grade polystyrene
  • All dishes sterilized using e-beam sterilization
  • Convenient packaging in easy open sleeves
  • 25 Sleeves/Cs.
Specifications
Specifications
Stackable
Bi Dish, I-Plate
24/Sleeve, 25 Sleeves/Cs.
Polystyrene
Raised Ridge
15 mm
Videos
SDS
Documents
Documents
Accessories