Fisherbrand™ Compartmentalized Petri Dishes
Select from 2, 3, or 4-section plates with numbered sections
 View more versions of this product
Catalog No. FB087583
Explore available promotions
Description
Description
Select from 2, 3, or 4-section plates with numbered sections
  • All dishes manufactured from virgin medical grade polystyrene
  • All dishes sterilized using e-beam sterilization
  • Convenient packaging in easy open sleeves
  • 25 Sleeves/Cs.
Specifications
Specifications
Stackable
Sterile
Raised Ridge
15 mm
Polystyrene
Tri Dish, Y-Plate
24/Sleeve, 25 Sleeves/Cs.
65 mm