Learn More
Fisherbrand™ Cryogenic Vial Storage Boxes
Suitable for ultra-low temperature storage of both internally and externally threaded cryogenic vials
Supplier:  Fisherbrand™ 3004630909
 View more versions of this product
Catalog No. 10-500-29
Only null left
Explore available promotions
Description
Description
Suitable for ultra-low temperature storage of both internally and externally threaded cryogenic vials
  • 81-Place white polycarbonate storage boxes
  • Use in mechanical freezers and liquid nitrogen Dewars
  • Cover is printed with a 9 x 9 numbered grid for inventory purposes
  • Stackable
  • Autoclavable
Specifications
Specifications
White
Square
For ultra-low temperature storage of both internally and externally threaded cyrogenic vials
9 x 9
5.25 in.
5.25 in.
3.75 in.
Polycarbonate
Cryo Storage Box
5 mL Cryo Vials
5.25 x 5.25 x 3.75 in. (13.3 x 13.3 x 9.5 cm)
13.3 cm
13.3 cm
9.5 cm
Product Suggestions
Product Suggestions