Fisherbrand™ Data-Coded Hollow Cathode 1 1/2in. dia. Lamps for Varian™ Instruments
For Varian™ instruments
 View more versions of this product
Catalog No. 14-386-115N
Explore available promotions
Description
Description
Specifications
Specifications
Spectrophotometer Lamp
Zinc
Hollow Cathode
213.9 nm
1.5 in.
Varian™ instruments
0.5 in.
1.4 cm
Y-Shape
4 Pin Octal
Neon
500 hr.
10 mA
3.7 cm
5 in.
15.1 cm
Quartz