Fisherbrand™ Disposable Aluminum Weighing/Drying Pans
Designed for weighing, drying and more. Compatible with many popular balances and moisture analyzers. Choice of 10.9cm and 12.2cm diameter dishes.
 View more versions of this product
Catalog No. 087-32-111
Explore available promotions
Description
Description
Fisherbrand™ Disposable Aluminum Weighing/Drying Pans are an economical and reliable choice for single-use lab applications. Designed for weighing, drying and more. Compatible with many popular balances and moisture analyzers. Choice of 10.9cm and 12.2cm diameter dishes.
  • General-purpose weighing pans also used for dispensing, storage, evaporating and drying
  • Compatible with many popular balances and moisture analyzers, including:
    • Ohaus™, Mettler Toledo™, Sartorius™, Denver Instrument™ and more
  • Constructed of durable aluminum to hold up to lab uses
  • Choice of:
    • 10.9cm (4.25 in.) dishes with slightly fluted sides and beaded rim
    • 12.2cm (4.75 in.) dishes with flat tops
Specifications
Specifications
Flat
Single-use
Circular
General purpose
100.5 mm
60 mL
Aluminum
Weighing Pan
8 mm
100.5 mm (Inner Top)