Fisherbrand™ Disposable Borosilicate Glass Tubes with Polypropylene Screw Cap
Provides chemical stability and biological inertness of hard, borosilicate glass at price of disposable tubes
 View more versions of this product
Catalog No. 14-962-26M
Explore available promotions
Description
Description
Provides chemical stability and biological inertness of hard, borosilicate glass at price of disposable tubes
  • Natural linerless polypropylene screw cap
  • With marking spot
Ideal for tissue culture, bacteriology, clinical chemistry, blood typing, and crossmatching procedures
Specifications
Specifications
30 mL
Borosilicate Glass
Provides chemical stability and biological inertness of hard, borosilicate glass at price of disposable tubes
Tray
Non-sterile
Screw Cap
150 mm
Single-use
Marking Spot
250/Tray
20 mm
Disposable Culture Tube