missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Disposable Petri Dishes
Tight-fitting lid prevents dehydration.
Description
Secure, tight-fitting lid helps to prevent dehydration.
Sterilized Petri Dishes
- 47mm dishes can be used with solid agar medium or add absorbent pads for liquid medium
- Molded polystyrene
Specifications
Single-use | |
Round | |
Petri Dish without Pad | |
47 mm |
Polystyrene | |
Sterile | |
150 Pack |