missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Disposable Petri Dishes
Tight-fitting lid prevents dehydration.
$91.14 - $302.09
Specifications
Disposable | Single-use |
---|---|
Material | Polystyrene |
Sterility | Sterile |
Diameter (Metric) | 47 mm |
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
09-720-500
|
N/A | Petri Dish without Pad | 150 Pack |
Pack of 150 Each for $91.14
|
|
||||
09-720-501
|
N/A | Petri Dish with Pad | 150 Pack |
Pack of 150 Each for $102.56
|
|
||||
09-720-502
|
N/A | Petri Dish without Pad | 600 Pack |
Pack of 600 Each for $302.09
|
|
||||
09-720-503
|
N/A | Petri Dish with Pad | 600 Pack |
Pack of 600 Each for $277.31
|
|
||||
Description
Secure, tight-fitting lid helps to prevent dehydration.
Available with or without absorbant pads
Sterilized Petri Dishes
- 47mm dishes can be used with solid agar medium or add absorbent pads for liquid medium
- Molded polystyrene
- Preloaded 47mm dishes with sterilized absorbent pads
- Minimize contamination
- Pure cellulose pad
- Extended 3-year shelf-life
Specifications
Single-use | |
Polystyrene | |
Sterile | |
47 mm |