Fisherbrand™ Disposable Petri Dishes
Tight-fitting lid prevents dehydration.
 View more versions of this product
Catalog No. 09-720-503
Explore available promotions
Description
Description
Secure, tight-fitting lid helps to prevent dehydration.
Sterilized Petri Dishes
  • 47mm dishes can be used with solid agar medium or add absorbent pads for liquid medium
  • Molded polystyrene
Sterilized Pad Dishes
  • Preloaded 47mm dishes with sterilized absorbent pads
  • Minimize contamination
  • Pure cellulose pad
  • Extended 3-year shelf-life
Specifications
Specifications
Single-use
Round
Petri Dish with Pad
47 mm
Polystyrene
Sterile
600 Pack