Fisherbrand™ Dissecting Jewelers Microforceps
For very precise work
 View more versions of this product
Catalog No. 08-953F
Explore available promotions
Description
Description
For very precise work
  • Fisherbrand™ Dissecting Jewelers Microforceps are manufactured to exact standards and exceed perfromance expectations
  • Ultrathin, smooth tips for the finest precision work
  • Smooth handles for comfort and ease of use
  • Passivated for corrosion resistance
  • ASTM™ standard stainless steel construction
  • Latex-free
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully autoclavable
Specifications
Specifications
Dissecting Forcep
ASTM, FDA
Yes
4.5 in.
Autoclavable
Stainless Steel
Curved, smooth
11.4 cm