Fisherbrand™ Electroporation Cuvettes Plus™
Disposable, color-coded cuvets fit most electroporators. Sterilized by gamma irradiation and supplied individually wrapped.
 View more versions of this product
Catalog No. FB101
Explore available promotions
Description
Description
Disposable, color-coded cuvets are compatible with most electroporators.
  • Clear, medical grade polycarbonate for high visibility
  • Polished aluminum lowers arcing frequency
  • Round cap allows single-handed removal
  • FB102 has imbedded aluminum electrodes
Specifications
Specifications
Sterile
90 μL
Imbedded aluminum electrodes
Electroporator
Electroporation Cuvet
Polycarbonate
1 mm