Fisherbrand™ Electroporation Cuvettes Plus™
Disposable, color-coded cuvets fit most electroporators. Sterilized by gamma irradiation and supplied individually wrapped.
$680.07 - $722.15
Specifications
Sterility Sterile
Type Electroporation Cuvet
Material Polycarbonate
Includes Imbedded aluminum electrodes
For Use With (Equipment) Electroporator
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Width (Metric) Gap Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Width (Metric) Gap Price Quantity  
FB101
View Documents
N/A 90 μL 1 mm
Pack of 50 Each for $680.07
Case of 50 Pack for $34,003.50
FB102
View Documents
N/A 400 μL 2 mm
Pack of 50 Each for $722.15
Case of 50 Pack for $36,107.50
FB104
View Documents
N/A 800 μL 4 mm
Pack of 50 Each for $722.15
Case of 50 Pack for $36,107.50
Description
Description
Disposable, color-coded cuvets are compatible with most electroporators.
  • Clear, medical grade polycarbonate for high visibility
  • Polished aluminum lowers arcing frequency
  • Round cap allows single-handed removal
  • FB102 has imbedded aluminum electrodes
Specifications
Specifications
Sterile
Polycarbonate
Electroporator
Electroporation Cuvet
Imbedded aluminum electrodes
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'