Fisherbrand™ Elite™ Adjustable-Volume Pipettes
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
Supplier:  Fisherbrand™ 4680000
 View more versions of this product
Catalog No. FBE00002
Explore available promotions
Description
Description
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
  • Fully autoclavable — entire pipette can be steam autoclaved
  • Soft-touch tip ejection — a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle — comfortable grip and effortless tip attachment
  • Optimal fit with SureOne™ pipette tips — best pipetting performance
  • Super blow-out piston — creates 150% bigger air push than conventional piston for delivery of the micro size drops
Specifications
Specifications
2.5 to 12%
Autoclavable
Elite™
0.2 to 2 μL
Calibration certificate, in-lab calibration tool
Soft touch tip ejection
SureOne™ tips
SureOne™ Tip
Micro Pipette
Ergonomic
0.002μL
3 Years
SDS
Documents
Documents
Accessories