Fisherbrand™ Elite™ Adjustable-Volume Pipettes
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
 View more versions of this product
Catalog No. FBE00200
Explore available promotions
Description
Description
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
  • Fully autoclavable — entire pipette can be steam autoclaved
  • Soft-touch tip ejection — a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle — comfortable grip and effortless tip attachment
  • Optimal fit with SureOne™ pipette tips — best pipetting performance
  • Super blow-out piston — creates 150% bigger air push than conventional piston for delivery of the micro size drops
Specifications
Specifications
0.6 to 1.8%
Autoclavable
Elite™
20 to 200 μL
Calibration certificate, in-lab calibration tool
Soft touch tip ejection
SureOne™ tips
SureOne™ Tip
Micro Pipette
Ergonomic
0.2μL
3 Years
SDS
Documents
Documents
Accessories