Learn More
Fisherbrand™ Elite™ Adjustable-Volume Pipettes
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
 View more versions of this product
Catalog No. FBE05000
Only null left
Explore available promotions
Description
Description
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
  • Fully autoclavable — entire pipette can be steam autoclaved
  • Soft-touch tip ejection — a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle — comfortable grip and effortless tip attachment
  • Super blow-out piston — creates 150% bigger air push than conventional piston for delivery of the micro size drops
Specifications
Specifications
0.5 to 2%
Elite
0.5 to 5 mL
Calibration certificate, in-lab calibration tool
Soft touch tip ejection
Autoclavable
Standard Pipette
Ergonomic
0.01mL
3 Years
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories
Promotions
Promotions
As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.