Fisherbrand™ Elite™ Variable Volume, Multichannel Pipettes
New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort
Supplier:  Fisherbrand™ 4682040
 View more versions of this product
Catalog No. FBE1200010
Explore available promotions
Description
Description
New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort
  • Fully autoclavable - entire pipetter can be steam autoclaved
  • Extremely low plunger forces - work longer without fatigue and reduce the risk of RSI
  • Soft-touch tip ejection—a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle - comfortable grip and effortless tip attachment
  • Optimal fit with SureOne pipetter tips - best pipetting performance
  • Super blow-out piston - creates 150% bigger air push than conventional piston for delivery of the micro size drops
Specifications
Specifications
2.4 to 12%
Autoclavable
12
Variable Volume Pipette
Ergonomic
SureOne™ Pipetter tips
SureOne Pipette Tip
Elite
1 to 10 μL
0.02μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories