Fisherbrand™ Flexible-Handle Brushes Edge
Edge
Soft, single row of bristles fits easily into flask necks
 View more versions of this product
Catalog No. 03-570A
Edge
Explore available promotions
Description
Description
Designed for cleaning Erlenmeyer, filtering, Kjeldahl, Nephelo, pear-shaped, round-bottom, flat-bottom, reaction and volumetric flasks.
  • Plastic handle bends to allow fitting of bristles to container shape
  • Handle has hanging ring
Specifications
Specifications
Flask Brush
Plastic
1.37 in.
250mL flasks
12 in.
30.5 cm
1
Soft
Flexible
3.5 cm
Hanging ring
3.5 in.
8.9 cm