missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Fluorescence Rectangular Cells: Macro Cells With PTFE Lids

Fit standard spectrophotometer cell holders

Supplier:  Fisherbrand™ 14958130

 View more versions of this product

Catalog No. 14-958-130



Explore available promotions

Description

Description

Fit standard spectrophotometer cell holders

 

  • All cells are 12.5mm wide
  • 10mm path length rectangular cells
  • Reduce reagent use with micro cells
  • Wavelength range for all cells is 200 to 2500nm
  • Quartz windows for transmission from far ultraviolet through the infrared

 

Specifications

Specifications

Macro Fluorescence Cell
Quartz
10mm
10 mm
45 mm
12.5 mm
Rectangle
3000 μL
PTFE
4/Pk., 4 Pk./Cs.
200 to 2500 nm
Standard Spectrophotometer Cell Holder
Transmission from far ultraviolet through the infrared
Videos
SDS
Documents

Documents

Promotions

Promotions