missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Fluorescence Rectangular Cells: Macro Cells With PTFE Lids
Fit standard spectrophotometer cell holders
$583.55 - $1172.93
Specifications
Type | Macro Fluorescence Cell |
---|---|
Capacity (Metric) | 3000 μL |
Material | Quartz |
Closure Material | PTFE |
Width (Metric) Interior | 10mm |
Includes: PTFE lid or stopper
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
14-958-128
|
Fisherbrand™
14958128 |
2/Pk., 8 Pk./Cs. |
Pack of 2 Each for $583.55
Case of 8 Pack for $4,668.40
|
|
|||||
14-958-130
|
Fisherbrand™
14958130 |
4/Pk., 4 Pk./Cs. |
Pack of 4 Each for $1,172.93
Case of 4 Pack for $4,691.72
|
|
|||||
Description
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 10mm path length rectangular cells
- Reduce reagent use with micro cells
- Wavelength range for all cells is 200 to 2500nm
- Quartz windows for transmission from far ultraviolet through the infrared
Specifications
Macro Fluorescence Cell | |
Quartz | |
10mm | |
200 to 2500 nm | |
Standard Spectrophotometer Cell Holder | |
Transmission from far ultraviolet through the infrared |
3000 μL | |
PTFE | |
10 mm | |
45 mm | |
12.5 mm | |
Rectangle |