Fisherbrand™ Free-Standing Microcentrifuge Tubes with Screw Caps
Autoclavable polypropylene
 View more versions of this product
Catalog No. 02-682-557
Explore available promotions
Description
Description
Autoclavable polypropylene
  • Withstand a maximum force of 20,000 × G
  • Free-standing with cap
  • Natural color
Order Info
500 per pack, 10 packs per case
Specifications
Specifications
Polypropylene
1.5 mL
Polypropylene
Yes
20,000 x g
Autoclavable
Natural
Non-sterile
Screw Cap