Fisherbrand™ Free-Standing Microcentrifuge Tubes with Screw Caps
Autoclavable polypropylene
$284.61 - $411.42
Specifications
Closure Material Polypropylene
Autoclavable Autoclavable
Color Natural
Material Polypropylene
Sterility Non-sterile
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
02-682-557
View Documents
N/A 1.5 mL
Pack of 500 Each for $411.42
Case of 10 Pack for $4,114.20
02-682-558
View Documents
Fisherbrand™
520GRDFIS
2 mL
Pack of 500 Each for $284.61
Case of 10 Pack for $2,846.10
02-682-559
View Documents
N/A 0.5 mL
Pack of 500 Each for $409.88
Case of 10 Pack for $4,098.80
Description
Description
Autoclavable polypropylene
  • Withstand a maximum force of 20,000 × G
  • Free-standing with cap
  • Natural color
Order Info
500 per pack, 10 packs per case
Specifications
Specifications
Polypropylene
Natural
Non-sterile
20,000 x g
Autoclavable
Polypropylene
Screw Cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'