Fisherbrand™ General Filtration Membrane Filters Edge
Edge
Useful for most filter applications
 View more versions of this product
Catalog No. 09-719-2B
Edge
Description
Description
  • Plain, white membrane filters
  • Pure, biologically inert mixture of cellulose acetate and cellulose nitrate (MCE)
  • Compatible with dilute acids and bases, aliphatic and aromatic hydrocarbons, and nonpolar liquids
  • Filters wet readily with water and other fluids
  • Contain no fibers or particles to contaminate filtrate
  • Autoclavable
Applications:
For all analytical, ultracleaning, and sterilizing applications at temperatures below 75°C (167°F)—except those involving esters, ether-alcohols, ketones, nitroparaffins or strong acids and bases
Specifications
Specifications
Autoclavable
0.22 μm
47 mm
MCE
For all analytical, ultracleaning, and sterilizing applications at temperatures below 75°C (167°F) – except those involving esters, ether-alcohols, ketones, nitroparaffins or strong acids and bases.