Fisherbrand™ Graduated Cylinders
Single metric scale in tough ceramic enamel.
Supplier:  Fisherbrand™ S63461
 View more versions of this product
Catalog No. S63461
Explore available promotions
Description
Description
  • Quality borosilicate glass cylinder with pouring lip
  • Heavy glass base provides extra stability
Specifications
Specifications
Round
Round
To Contain
1000 mL
10 mL
±6.0 mL
±6 mL
Graduated Cylinder
Borosilicate Glass
Single
Autoclavable
10 mL