Fisherbrand™ Histosette™ II Tissue Processing/Embedding Cassettes
The Histosette™ II Tissue Processing/Embedding Cassettes have an anterior writing area is slanted at a 45° angle to make the cassette more suitable to be used with certain types of cassette labeling instruments.
Supplier: Fisherbrand™ M4858FS
Description
- Made of acetal
- Unobstructed writing area at a 45° angle
- Cassettes and lids packaged separately
- Front hinge design
- A secure locking device safely holds the specimen during processing
Specifications
Tan | |
Tissue Cassette |