Fisherbrand™ Histosette™ II Tissue Processing/Embedding Cassettes
The Histosette™ II Tissue Processing/Embedding Cassettes have an anterior writing area is slanted at a 45° angle to make the cassette more suitable to be used with certain types of cassette labeling instruments.
$219.87 - $219.87
Specifications
Product Type Tissue Cassette
View More Specs
Products 2
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
15-182-708F
View Documents
Fisherbrand™
M4857FS
Peach
Case of 1000 Each for $219.87
15-182-708G
View Documents
Fisherbrand™
M4858FS
Tan
Case of 1000 Each for $219.87
Description
Description
  • Made of acetal
  • Unobstructed writing area at a 45° angle
  • Cassettes and lids packaged separately
  • Front hinge design
  • A secure locking device safely holds the specimen during processing
Specifications
Specifications
Tissue Cassette
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'