Fisherbrand™ Hollow Cathode Single-Element 2in. dia. Lamps
Window materials are quartz, borosilicate and UV glass; Lamps are single-element and 2 in. diameter
$869.67 - $1201.87
Catalog Number | Mfr. No. | Amperage | Element | Material | Wavelength | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Amperage | Element | Material | Wavelength | Price | Quantity | ||
14-386-104R
|
N/A | 18 mA | Arsenic | Quartz | 193.7 nm |
Each for $931.63
|
|
||
14-386-105S
|
N/A | 30 mA | Iron | UV Glass | 248.3 nm |
Each for $869.67
|
|
||
14-386-105A
|
N/A | 12 mA | Chromium | UV Glass | 357.9 nm |
Each for $929.39
|
|
||
14-386-105AA
|
N/A | 15 mA | Lead | Quartz | 217 nm |
Each for $903.26
|
|
||
14-386-105Y
|
N/A | 30 mA | Manganese | UV Glass | 279.5 nm |
Each for $992.85
|
|
||
14-386-106C
|
N/A | 30 mA | Nickel | Quartz | 232 nm |
Each for $955.52
|
|
||
14-386-106V
|
N/A | Sodium | UV Glass |
Each for $1,026.43
|
|
||||
14-386-107M
|
N/A | 20 mA | Zinc | Quartz | 213.9 nm |
Each for $1,201.87
|
|
||
Description
- Lamps are single-element and have 2 in. (51mm) diameter
- Direct replacement lamps for Perkin-Elmer™ nondata-coded instruments
- Argon or neon gas fills
- Window materials are quartz, borosilicate glass or UV glass