Fisherbrand™ L-Shaped Cell Spreaders Edge
Edge
Ideal for even spreading of bacterial culture
 View more versions of this product
Catalog No. 14-665-230
Edge
Explore available promotions
Description
Description
For spreading of bacterial culture on agar plates
  • Disposable
  • Smooth spreading surface with slight upward angle reduces the risk of damaging agar sample
  • L-shape allows application on the entire Petri dish or plate surface
  • Presterilized blue polypropylene eliminates the need for flaming or autoclaving
Specifications
Specifications
Single-use
L-Shape
Blue
Polypropylene
Ideal for even spreading of bacterial cultures, smooth spreading surface with slight upward angle reduces the risk of damaging agar sample
Sterile
Cell Spreader
10 each per pouch, 50 pouches/case
Agar Culture Plate