missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ L-Shaped Cell Spreaders
Ideal for even spreading of bacterial culture
$37.15 - $65.26
Specifications
Color | Blue |
---|---|
Material | Polypropylene |
Product Type | Cell Spreader |
Sterility | Sterile |
For Use With (Equipment) | Agar Culture Plate |
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity | ||||
14-665-230
|
N/A | Cell Spreader | 10 each per pouch, 50 pouches/case |
Pack of 500 Each for $65.26
|
|
||||
14-665-231
|
N/A | Cell Spreader | 1 each per pouch, 100 pouches/case |
Pack of 100 Each for $37.15
|
|
||||
Description
For spreading of bacterial culture on agar plates
- Disposable
- Smooth spreading surface with slight upward angle reduces the risk of damaging agar sample
- L-shape allows application on the entire Petri dish or plate surface
- Presterilized blue polypropylene eliminates the need for flaming or autoclaving
Specifications
Blue | |
Cell Spreader | |
Agar Culture Plate | |
Single-use |
Polypropylene | |
Sterile | |
L-Shape | |
Ideal for even spreading of bacterial cultures, smooth spreading surface with slight upward angle reduces the risk of damaging agar sample |