Fisherbrand™ Lab-Label Adhesive Lab Labels
Won't stick to latex or vinyl gloves.
Description
- Ideal for use in blood banks and hospitals—anywhere that gloves are used
- Permanent adhesive adheres to test tubes and other surfaces
- Labels suited for use in freezers and water baths
- Peel-off labels do not leave residue on tubes or gloves
- Smudge-resistant
- Easy to reposition
Specifications
Permanent | |
Vinyl | |
Ideal for use in blood banks and hospitals anywhere that gloves are used | |
3 in. | |
1.93 in. |
Label | |
Adhesive Lab Label | |
500/Pk. | |
7.62 cm |