Fisherbrand™ Lab-Label Adhesive Lab Labels
Won't stick to latex or vinyl gloves.
 View more versions of this product
Catalog No. 15924
Explore available promotions
Description
Description
  • Ideal for use in blood banks and hospitals—anywhere that gloves are used
  • Permanent adhesive adheres to test tubes and other surfaces
  • Labels suited for use in freezers and water baths
  • Peel-off labels do not leave residue on tubes or gloves
  • Smudge-resistant
  • Easy to reposition
Specifications
Specifications
Permanent
Vinyl
Ideal for use in blood banks and hospitals anywhere that gloves are used
3 in.
0.75 in.
Label
Adhesive Lab Label
500/Pk.
7.62 cm